Stampa…

5-sulphoanthranilic acid ≥96.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Amino-5-sulphobenzoic acid

S0802-25G S0802-5G
TCIAS0802-25GEA 249 EUR
TCIAS0802-25G TCIAS0802-5G
5-sulphoanthranilic acid ≥96.0% (by HPLC, titration analysis)
5-sulphoanthranilic acid
Formula: C₇H₇NO₅S
Peso molecolare: 217,20 g/mol
Punto di fusione: 317°C
MDL Number: MFCD00035763
CAS Number: 3577-63-7
EINECS: 222-697-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Neutralization titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR