Stampa…

Salbutamol sulphate ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: Bronchospray, Proventil-HFA, Salbutamol hemisulphate, Albuterol hemissulphate, 2-(tert-Butylamino)-1-(4-hydroxy-3-hydroxymethylphenyl)ethanol sulphate, Albutarol sulphate, 5-[2-(tert-Butylamino)-1-hydroxyethyl]-2-hydroxybenzyl Alcohol sulphate

S0531-25G S0531-5G S0531-1G
TCIAS0531-25GEA 269 EUR
TCIAS0531-25G TCIAS0531-5G TCIAS0531-1G
Salbutamol sulphate ≥98.0% (by HPLC, titration analysis)
Salbutamol sulphate
Formula: (C₁₃H₂₁NO₃)₂·H₂SO₄
Peso molecolare: 576,702 g/mol
Punto di fusione: 180°C
MDL Number: MFCD00055200
CAS Number: 51022-70-9
EINECS: 256-916-8
Merck Index: 14,00216
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa3e7f0dc1db6555cd2e899bc45d5a7008fe93c3&vid=fa3e7f0dc1db6555cd2e899bc45d5a7008fe93c3&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR