Stampa…

Sertraline hydrochloride ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI
Attenzione

S0507-1G S0507-5G
TCIAS0507-1GEA 18 EUR
TCIAS0507-1G TCIAS0507-5G
Sertraline hydrochloride ≥98.0% (by HPLC, titration analysis)
Sertraline hydrochloride
Formula: C₁₇H₁₇Cl₂N
Punto di fusione: 245°C
MDL Number: MFCD00895772
CAS Number: 79559-97-0
Merck Index: 14,08467

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 245.0 to 249.0 deg-C
Specific rotation [a]20/D +35.0 to +41.0 deg (C=2,MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR