Stampa…

ɑ-Sexithiophene, purified by sublimation

Fornitore: TCI

Synonyms: 6T

S0504-1G S0504-100MG
TCIAS0504-1GEA 138 EUR
TCIAS0504-1G TCIAS0504-100MG
ɑ-Sexithiophene, purified by sublimation
alpha-Sexithiophene
Formula: C₂₄H₁₄S₆
Peso molecolare: 494.74 g/mol
Punto di fusione: 304°C
CAS Number: 88493-55-4

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
TOF-MS to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR