Stampa…

4-Sulphothiacalix[4]arene sodium salt ≥98.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: Tetrasodium Tetrathiacalix[4]arenetetrasulphonate, Thiacalix[4]arenetetrasulphonic acid Tetrasodium Salt

S0477-1G S0477-5G
TCIAS0477-1GEA 79 EUR
TCIAS0477-1G TCIAS0477-5G
4-Sulphothiacalix[4]arene sodium salt ≥98.0% (by HPLC)
4-Sulphothiacalix[4]arene sodium salt
Formula: C₂₄H₁₆O₁₆S₈
Peso molecolare: 904,78 g/mol
Punto di fusione: 390°C
MDL Number: MFCD06797166
CAS Number: 211561-04-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR