Stampa…

4-Sulphocalix[4]arene hydrate ≥94.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Tetrahydroxycalix[4]arenetetrasulphonic acid, Tetrasulpho(tetrahydroxy)calix[4]arene

S0469-1G S0469-5G
TCIAS0469-1GEA 174 EUR
TCIAS0469-1G TCIAS0469-5G
4-Sulphocalix[4]arene hydrate ≥94.0% (by titrimetric analysis)
4-Sulphocalix[4]arene hydrate
Formula: C₂₈H₂₄O₁₆S₄
Peso molecolare: 744,72 g/mol
MDL Number: MFCD03095486
CAS Number: 112269-92-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Gray to Red powder to crystal
Purity (Neutralization titration) min. 94.0 % (calcd.on anh.substance)
Water max. 15.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR