Stampa…

Disodium sebacate ≥97.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: Sodium decanedioate, Decanedioic Acid Disodium Salt, Disodium Decanedioate, Disodium Sebacate, Sebacic Acid Disodium Salt

S0270-25G S0270-500G
TCIAS0270-25GEA 49 EUR
TCIAS0270-25G TCIAS0270-500G
Disodium sebacate ≥97.0% (by titrimetric analysis)
Disodium sebacate
Formula: C₁₀H₁₈O₄
Peso molecolare: 246,21 g/mol
MDL Number: MFCD00070501
CAS Number: 17265-14-4
EINECS: 241-300-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f789f9297d4ac7073531c76c01294c4521ab3247&vid=f789f9297d4ac7073531c76c01294c4521ab3247&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR