Diisopropyl succinate ≥99.0% (by GC)
Fornitore: TCI
S0269-25ML
S0269-500ML
TCIAS0269-25MLEA
22
EUR
TCIAS0269-25ML
TCIAS0269-500ML
Diisopropyl succinate ≥99.0% (by GC)
Diisopropyl succinate
Formula:
C₁₀H₁₈O₄ Punto di ebollizione: 228°C |
MDL Number:
MFCD00059360 CAS Number: 924-88-9 EINECS: 213-110-0 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...