Stampa…

2-Sulphobenzaldehyde sodium salt ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: 2-Formylbenzenesulphonic acid sodium salt, Sodium 2-formylbenzenesulphonate, Sodium 2-Formylbenzenesulphonate

S0122-25G S0122-500G
TCIAS0122-25GEA 43 EUR
TCIAS0122-25G TCIAS0122-500G
2-Sulphobenzaldehyde sodium salt ≥98.0% (by HPLC, titration analysis)
2-Sulphobenzaldehyde sodium salt
Formula: C₇H₆O₄S
MDL Number: MFCD00007478
CAS Number: 1008-72-6
EINECS: 213-758-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Ion exchange titration) min. 98.0 % (calcd.on anh.substance)
Water max. 5.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR