Stampa…

Sodium sulphamate ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: sulphamic acid Sodium Salt, Aminosulphonic acid Sodium Salt

S0118-25G S0118-500G
TCIAS0118-25GEA 22 EUR
TCIAS0118-25G TCIAS0118-500G
Sodium sulphamate ≥98.0% (by titrimetric analysis)
Sodium sulphamate
Formula: H₃NO₃S
Peso molecolare: 119,07 g/mol
MDL Number: MFCD00059940
CAS Number: 13845-18-6
EINECS: 237-572-8

Visualizza i codici

Risultati del test di specificazione

Appearance White powder to crystal
Purity (Ion exchange titration) min. 98.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR