Stampa…

Stigmasterol ≥90.0% (by GC)

Fornitore: TCI EUROPE

S0088-1G S0088-25G S0088-5G
TCIAS0088-1GEA 21 EUR
TCIAS0088-1G TCIAS0088-25G TCIAS0088-5G
Stigmasterol ≥90.0% (by GC)
Stigmasterol
Formula: C₂₉H₄₈O
Peso molecolare: 412,70 g/mol
MDL Number: MFCD00003630
CAS Number: 83-48-7
EINECS: 201-482-7
Merck Index: 14,08815
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=218a94dfd7d4acf8fb0f9cd6663e0aa243ac7267&vid=218a94dfd7d4acf8fb0f9cd6663e0aa243ac7267&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 90.0 %
Melting point 166.0 to 170.0 deg-C
Specific rotation [a]20/D -48.0 to -53.0 deg (C=2, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR