Stampa…

Spermine phosphate hexahydrate ≥98.0% (by total nitrogen basis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Neuridine Phosphate, Gerontine Phosphate, N,N'-Bis(3-aminopropyl)tetramethylenediamine Phosphate, Musculamine Phosphate, N,N'-Bis(3-aminopropyl)-1,4-butanediamine Phosphate

S0069-1G S0069-5G
TCIAS0069-1GEA 47 EUR
TCIAS0069-1G TCIAS0069-5G
Spermine phosphate hexahydrate ≥98.0% (by total nitrogen basis)
Spermine phosphate hexahydrate
Formula: C₁₀H₂₆N₄
Peso molecolare: 398,33 g/mol
MDL Number: MFCD00050839
CAS Number: 58298-97-8
EINECS: 223-441-2
Merck Index: 14,08743

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 % (calcd.on anh.substance)
Solubility in dilute HCl almost transparency
Water 20.0 to 23.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR