Stampa…

Serotonin creatinine sulphate monohydrate ≥98.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

Synonyms: 5-Hydroxytryptamine Creatinine Sulphate

S0036-1G S0036-100MG
TCIAS0036-1GEA 53 EUR
TCIAS0036-1G TCIAS0036-100MG
Serotonin creatinine sulphate monohydrate ≥98.0% (by titrimetric analysis)
Serotonin creatinine sulphate monohydrate
Formula: C₁₀H₁₂N₂O
Peso molecolare: 387.41 g/mol
MDL Number: MFCD00149653
CAS Number: 61-47-2
Merck Index: 14,08462

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) min. 98.0 %
Water 4.0 to 4.7 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR