Stampa…

Ro 5-3335 ≥98.0% (by HPLC)

Fornitore: TCI

R0223-10MG R0223-50MG
TCIAR0223-10MGEA 147 EUR
TCIAR0223-10MG TCIAR0223-50MG
Ro 5-3335 ≥98.0% (by HPLC)
Ro 5-3335
Formula: C₁₃H₁₀ClN₃O
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00870704
CAS Number: 30195-30-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light red to Green powder to crystal
Purity (HPLC) min. 98.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR