RS-127445 hydrochloride ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
RS-127445 HCl, 4-(4-Fluoronaphthalen-1-yl)-6-isopropylpyrimidin-2-amine hydrochloride
R0221-10MG
R0221-50MG
TCIAR0221-10MGEA
136
EUR
TCIAR0221-10MG
TCIAR0221-50MG
RS-127445 hydrochloride ≥98.0% (by HPLC)
RS-127445 hydrochloride
Formula:
C₁₇H₁₆FN₃·HCl Peso molecolare: 317,79 g/mol Punto di fusione: 158°C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD11112196 CAS Number: 199864-86-3 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Nitrogen) | 12.60 to 13.80 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...