Stampa…

Rotigotine ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: (S)-6-[Propyl[2-(thiophen-2-yl)ethyl]amino]-5,6,7,8-tetrahydronaphthalen-1-ol

R0220-10MG R0220-50MG
TCIAR0220-10MGEA 113 EUR
TCIAR0220-10MG TCIAR0220-50MG
Rotigotine ≥98.0% (by HPLC, titration analysis)
Rotigotine
Formula: C₁₉H₂₅NOS
Peso molecolare: 315,48 g/mol
Punto di fusione: 78°C
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00869590
CAS Number: 99755-59-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -34.0 ~ -38.0 deg (C=1, methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR