Ramelteon ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
(S)-N-(2-(1,6,7,8-Tetrahydro-2H-indeno[5,4-b]furan-8-yl)ethyl)propionamide, (S)-N-[2-(2,6,7,8-Tetrahydro-1H-indeno[5,4-b]furan-8-yl)ethyl]propionamide
R0216-100MG
R0216-1G
TCIAR0216-100MGEA
68
EUR
TCIAR0216-100MG
TCIAR0216-1G
Ramelteon ≥98.0% (by GC)
Ramelteon
Formula:
C₁₆H₂₁NO₂ Peso molecolare: 259,35 g/mol |
MDL Number:
MFCD08067736 CAS Number: 196597-26-9 Merck Index: 14,08101 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 114.0 to 118.0 deg-C |
Specific rotation [a]20/D | -55.0 to -60.0 deg (C=1, CHCL3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...