Raltitrexed ≥98.0% (by HPLC)
Fornitore: TCI
|
Pericolo
|
R0208-50MG
R0208-250MG
TCIAR0208-50MGEA
146
EUR
TCIAR0208-50MG
TCIAR0208-250MG
Raltitrexed ≥98.0% (by HPLC)
Raltitrexed
Formula:
C₂₁H₂₂N₄O₆S Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD00864168 CAS Number: 112887-68-0 UN: 2811 ADR: 6.1,III Merck Index: 14,08099 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 97.0 % |
Specific rotation [a]20/D | -15.0 to -20.0 deg (C=1,MeOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...