RO-3306 ≥95.0% (by HPLC)
Fornitore: TCI EUROPE
Synonyms:
(Z)-5-(Quinolin-6-ylmethylene)-2-((thiophen-2-ylmethyl)amino)thiazol-4(5H)-one, (Z)-5-(Quinolin-6-ylmethylene)-2-[(thiophen-2-ylmethyl)amino]thiazol-4(5H)-one
R0201-10MG
R0201-50MG
TCIAR0201-10MGEA
144
EUR
TCIAR0201-10MG
TCIAR0201-50MG
RO-3306 ≥95.0% (by HPLC)
RO-3306
Formula:
C₁₈H₁₃N₃OS₂ Peso molecolare: 351,5 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD17392573 CAS Number: 872573-93-8 |
Risultati del test di specificazione
Appearance | White to Gray to Brown powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Elemental analysis (Nitrogen) | 11.00 to 13.00 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...