Stampa…
Rifapentine ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
Attenzione
Synonyms: (2S,12Z,14E,16S,17S,18R,19R,20R,21S,22R,23S,24E)-3-(((4-Cyclopentyl-1-piperazinyl)imino)methyl)-21-(Acetyloxy)-5,6,9,17,19-pentahydroxy-23-methoxy-2,4,12,16,18,20,22-heptamethyl-2,7-(epoxypentadeca[1,11,13]trienimino)naphtho[2,1-b]furan-1,11(2H)-dione
R0198-500MG R0198-100MG
TCIAR0198-500MGEA 365 EUR
TCIAR0198-500MG TCIAR0198-100MG
Rifapentine ≥98.0% (by HPLC)
Rifapentine
Formula: C₄₇H₆₄N₄O₁₂
Peso molecolare: 877,04 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00866806
CAS Number: 61379-65-5
EINECS: 262-743-9
Merck Index: 14,08219
Visualizza i codici
Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 97.0 %