Stampa…

Ru(II)-(S)-Pheox for use as a catalyst

Fornitore: TCI EUROPE
Attenzione

Synonyms: Tetrakis(acetonitrile)[2-[(4S)-4,5-dihydro-4-phenyl-2-oxazolyl-N]phenyl]ruthenium(II) Hexafluorophosphate, Ruthenium-tetrakis(acetonitrile)[2-[(4S)-4,5-dihydro-4-phenyl-2-oxazolyl-κN3]phenyl-κC]-hexafluorophosphate

R0196-200MG R0196-1G
TCIAR0196-200MGEA 70 EUR
TCIAR0196-200MG TCIAR0196-1G
Ru(II)-(S)-Pheox for use as a catalyst
Ru(II)-(S)-Pheox
Formula: C₂₃H₂₄F₆N₅OPRu
Peso molecolare: 632,51 g/mol
MDL Number: MFCD31618155
CAS Number: 1259070-80-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7aadba8886c85aa9b7233e2d7126238459e1664d&vid=7aadba8886c85aa9b7233e2d7126238459e1664d&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Amber to Dark green powder to crystal
Elemental analysis (Nitrogen) 10.50 to 11.50 %
Specific rotation [a]20/D +242 to +252 deg (C=0.1, CH3CN)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR