Roflumilast ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3-(Cyclopropylmethoxy)-N-(3,5-dichloropyridin-4-yl)-4-(difluoromethoxy)benzamide
R0193-1G
R0193-250MG
TCIAR0193-1GEA
182
EUR
TCIAR0193-1G
TCIAR0193-250MG
Roflumilast ≥98.0% (by HPLC)
Roflumilast
Formula:
C₁₇H₁₄Cl₂F₂N₂O₃ Peso molecolare: 403,21 g/mol Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD00938270 CAS Number: 162401-32-3 Merck Index: 14,08249 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Melting point | 157.0 to 161.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...