Stampa…

Rizatriptan benzoate ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI

Synonyms: N,N-Dimethyl-2-[5-(1,2,4-triazol-1-ylmethyl)-1H-indol-3-yl]ethylamine Benzoate

R0181-50MG R0181-250MG
TCIAR0181-50MGEA 73 EUR
TCIAR0181-50MG TCIAR0181-250MG
Rizatriptan benzoate ≥98.0% (by HPLC, titration analysis)
Rizatriptan benzoate
Formula: C₂₂H₂₅N₅O₂
Peso molecolare: 391.475 g/mol
MDL Number: MFCD00866224
CAS Number: 145202-66-0
Merck Index: 14,08241
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23c1596b3205d952545eddd2f9e263be9eaaa0e6&vid=23c1596b3205d952545eddd2f9e263be9eaaa0e6&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 % (total of Ion-Pair)
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 178.0 to 182.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR