Diacetato[(S)-(-)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl]ruthenium(II)
Fornitore: TCI
Synonyms:
Ru(OAc)2[(S)-tolbinap]
MDL Number:
MFCD09753021 CAS Number: 106681-15-6 |
|
Risultati del test di specificazione
Appearance | Dark green to Dark red to Black powder to crystal |
IR | to pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...