[RuCl(p-cymene)((R)-dtbm-segphos®)]Cl
Fornitore: TCI EUROPE
Synonyms:
Chloro{(R)-(-)-5,5'-bis[di(3,5-di-t-butyl-4-methoxyphenyl)phosphino]-4,4'-bi-1,3-benzodioxole}(p-cymene)ruthenium(II) chloride [RuCl(p-cymene)((R)-dtbm-segphos®)]Cl, Chloro[(R)-(-)-5,5'-bis[bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4'-bi-1,3-benzodioxole](p-cymene)ruthenium(II) Chloride
Formula:
C₈₄H₁₁₄Cl₂O₈P₂Ru Peso molecolare: 1485,75 g/mol |
MDL Number:
MFCD09753019 CAS Number: 944451-32-5 |
Risultati del test di specificazione
Appearance | Light yellow to Amber to Dark green powder to crystal |
IR | to pass test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...