Stampa…

[RuCl(p-cymene)((R)-dm-segphos®)]Cl

Fornitore: TCI EUROPE

Synonyms: Chloro[(R)-(+)-5,5'-bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole](p-cymene)ruthenium(II) Chloride, Chloro{(R)-(+)-5,5'-bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole}(p-cymene)ruthenium(II) chloride [RuCl(p-cymene)((R)-dm-segphos®)]Cl

R0156-200MG R0156-1G
TCIAR0156-200MGEA 44 EUR
TCIAR0156-200MG TCIAR0156-1G
[RuCl(p-cymene)((R)-dm-segphos®)]Cl
[RuCl(p-cymene)((R)-dm-segphos®)]Cl
Formula: C₅₆H₅₈Cl₂O₄P₂Ru
Peso molecolare: 1028,99 g/mol
MDL Number: MFCD09753018
CAS Number: 944451-30-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b95b690dc4af61bf720abaf729be7525bd989d50&vid=b95b690dc4af61bf720abaf729be7525bd989d50&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Amber to Dark green powder to crystal
IR to pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR