[RuCl(p-cymene)((S)-xylbinap)]Cl
Fornitore: TCI EUROPE
Synonyms:
Chloro{(S)-(-)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl}(p-cymene)ruthenium(II) chloride [RuCl(p-cymene)((S)-xylbinap)]Cl, Chloro{(S)-(-)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl}(p-cymene)ruthenium(II) chloride, Chloro[(S)-(-)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl](p-cymene)ruthenium(II) Chloride
Formula:
C₆₂H₆₂Cl₂P₂Ru Peso molecolare: 1041,10 g/mol |
MDL Number:
MFCD09753015 CAS Number: 944451-25-6 |
Risultati del test di specificazione
Appearance | Light yellow to Brown powder to crystal |
IR | to pass test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...