Stampa…

[RuCl(p-cymene)((R)-xylbinap)]Cl

Fornitore: TCI EUROPE

Synonyms: Chloro[(R)-(+)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl](p-cymene)ruthenium(II) Chloride, Chloro{(R)-(+)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl}(p-cymene)ruthenium(II) chloride [RuCl(p-cymene)((R)-xylbinap}]Cl

R0150-200MG R0150-1G
TCIAR0150-200MGEA 42 EUR
TCIAR0150-200MG TCIAR0150-1G
[RuCl(p-cymene)((R)-xylbinap)]Cl
[RuCl(p-cymene)((R)-xylbinap)]Cl
Formula: C₆₂H₆₂Cl₂P₂Ru
Peso molecolare: 1041,10 g/mol
MDL Number: MFCD09753015
CAS Number: 944451-24-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=437981766094f1614dd3ebd4113e6c71c8247cde&vid=437981766094f1614dd3ebd4113e6c71c8247cde&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
IR to pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR