Stampa…

[RuCl(p-cymene)((R)-tolbinap)]Cl

Fornitore: TCI EUROPE

Synonyms: Chloro[(R)-(+)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl](p-cymene)ruthenium(II) chloride

R0148-200MG R0148-1G
TCIAR0148-200MGEA 39 EUR
TCIAR0148-200MG TCIAR0148-1G
[RuCl(p-cymene)((R)-tolbinap)]Cl
[RuCl(p-cymene)((R)-tolbinap)]Cl
Formula: C₅₈H₅₄Cl₂P₂Ru
Peso molecolare: 984,99 g/mol
CAS Number: 1034001-51-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6385f8a5b040c44b779bff47b629abee7469a576&vid=6385f8a5b040c44b779bff47b629abee7469a576&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
IR to pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR