Stampa…

[RuCl(p-cymene)((R)-binap)]Cl ≥90.0% (by HPLC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: Chloro[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl](p-cymene)ruthenium(II) chloride [RuCl(p-cymene)((R)-binap)]Cl, (R)-RuCl[(p-cymene)(BINAP)]Cl, Chloro[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl](p-cymene)ruthenium(II) Chloride, [(R)-Binap RuCl p-cymene]Cl

R0146-200MG R0146-1G
TCIAR0146-200MGEA 36 EUR
TCIAR0146-200MG TCIAR0146-1G
[RuCl(p-cymene)((R)-binap)]Cl ≥90.0% (by HPLC)
[RuCl(p-cymene)((R)-binap)]Cl
Formula: C₅₄H₄₆Cl₂P₂Ru
Peso molecolare: 928,88 g/mol
MDL Number: MFCD00134456
CAS Number: 145926-28-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d9c9b3d85eb589b136423a752a33d12237c73e0&vid=6d9c9b3d85eb589b136423a752a33d12237c73e0&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 90 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR