Stampa…

RuCl2[(R)-xylbinap][(R,R)-dpen]

Fornitore: TCI EUROPE

Synonyms: Dichloro[(R)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl][(R,R)-1,2-diphenylethylenediamine]ruthenium(II), Dichloro{(R)-(+)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl}[(1R,2R)-(+)-1,2-diphenylethylenediamine]ruthenium(II) RuCl2[(R)-xylbinap][(R,R)-dpen]

R0135-200MG R0135-1G
TCIAR0135-200MGEA 39 EUR
TCIAR0135-200MG TCIAR0135-1G
RuCl2[(R)-xylbinap][(R,R)-dpen]
RuCl2[(R)-xylbinap][(R,R)-dpen]
Formula: C₆₆H₆₄Cl₂N₂P₂Ru
Peso molecolare: 1119,17 g/mol
MDL Number: MFCD09753028
CAS Number: 220114-38-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=94bdb4870db6a863dc73c03dcae76d8624ad59d3&vid=94bdb4870db6a863dc73c03dcae76d8624ad59d3&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Orange to Brown powder to crystal
IR to pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR