Stampa…

RuCl2[(S)-xylbinap][(S)-daipen]

Fornitore: TCI EUROPE
Pericolo

Synonyms: Dichloro{(S)-(-)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl}[(2S)-(+)-1,1-bis(4-methoxyphenyl)-3-methyl-1,2-butanediamine]ruthenium(II) RuCl2[(S)-xylbinap][(S)-daipen]

R0132-200MG R0132-1G
TCIAR0132-200MGEA 43 EUR
TCIAR0132-200MG TCIAR0132-1G
RuCl2[(S)-xylbinap][(S)-daipen]
RuCl2[(S)-xylbinap][(S)-daipen]
Formula: C₇₁H₇₄Cl₂N₂O₂P₂Ru
Peso molecolare: 1221,30 g/mol
MDL Number: MFCD09753027
CAS Number: 220114-01-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=74ea19a6434fb35dd5bf7acd0221e500bbcd9395&vid=74ea19a6434fb35dd5bf7acd0221e500bbcd9395&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Amber to Dark green powder to crystaline
IR to pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR