RuCl2[(R)-dm-segphos®][(R,R)-dpen]
Fornitore: TCI
Formula:
C₆₀H₆₀Cl₂N₂O₄P₂Ru |
MDL Number:
MFCD09753031 CAS Number: 944450-45-7 |
|
Risultati del test di specificazione
Appearance | Light yellow to Amber to Dark green powder to crystal |
IR | to pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...