RuCl2[(S)-dm-segphos®][(S)-daipen]
Fornitore: TCI
Formula:
C₆₅H₇₀Cl₂N₂O₆P₂Ru |
MDL Number:
MFCD09753029 CAS Number: 944450-44-6 |
|
Risultati del test di specificazione
Appearance | Light yellow to Amber to Dark green powder to crystal |
IR | to pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...