Stampa…
RuCl[(R,R)-Tsdpen](p-cymene) ≥90% (by HPLC)
Fornitore: TCI EUROPE
Synonyms: [(R,R)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulphonamide]chloro(p-cymene)ruthenium(II), RuCl(p-cymene)[(R,R)-Ts-DPEN], Chloro(p-cymene)[(R,R)-N-(p-toluenesulphonyl)-1,2-diphenylethylenediamine]ruthenium(II), Chloro{[(1R,2R)-(-)-2-amino-1,2-diphenylethyl](4-toluenesulphonyl)amido}(p-cymene)ruthenium(II)
R0125-200MG R0125-1G
TCIAR0125-200MGEA 36 EUR
TCIAR0125-200MG TCIAR0125-1G
RuCl[(R,R)-Tsdpen](p-cymene) ≥90% (by HPLC)
RuCl[(R,R)-Tsdpen](p-cymene)
Formula: C₃₁H₃₅ClN₂O₂RuS
Peso molecolare: 636,21 g/mol
MDL Number: MFCD10567385
CAS Number: 192139-92-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1f1dc31004440dc664187fb5515c120422364263&vid=1f1dc31004440dc664187fb5515c120422364263&prid=web&clid=SAPDAM
Visualizza i codici
Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 90 area%