Stampa…

Chloro{[(1S,2S)-(+)-2-amino-1,2-diphenylethyl](4-toluenesulphonyl)amido}(p-cymene)ruthenium(II) ≥90% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: Chloro(p-cymene)[(S,S)-N-(p-toluenesulphonyl)-1,2-diphenylethylenediamine]ruthenium(II), (S,S)-N-(p-Toluenesulphonyl)-1,2-diphenylethanediamine(chloro)(p-cymene)ruthenium(II), [(S,S)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulphonamide]chloro(p-cymene)ruthenium(II), RuCl[(S,S)-Tsdpen](p-cymene)

R0124-200MG R0124-1G
TCIAR0124-200MGEA 37 EUR
TCIAR0124-200MG TCIAR0124-1G
Chloro{[(1S,2S)-(+)-2-amino-1,2-diphenylethyl](4-toluenesulphonyl)amido}(p-cymene)ruthenium(II) ≥90% (by HPLC)
Chloro{[(1S,2S)-(+)-2-amino-1,2-diphenylethyl](4-toluenesulphonyl)amido}(p-cymene)ruthenium(II)
Formula: C₃₁H₃₅ClN₂O₂RuS
Peso molecolare: 636,21 g/mol
MDL Number: MFCD09953484
CAS Number: 192139-90-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83a865a06ef377080c334e52dcebbef8ec8ff684&vid=83a865a06ef377080c334e52dcebbef8ec8ff684&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 90 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR