RuCl[(R,R)-Fsdpen](p-cymene) ≥90% (by NMR)
Fornitore: TCI
Formula:
C₃₀H₂₈ClF₅N₂O₂RuS |
MDL Number:
MFCD12545963 CAS Number: 1026995-71-0 |
|
Risultati del test di specificazione
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (19F-qNMR) | min. 90 w/w% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...