Stampa…

RuCl[(S,S)-Fsdpen](p-cymene) ≥90% (by NMR)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Chloro(p-cymene)[(S,S)-N-(pentafluorobenzenesulphonyl)-1,2-diphenylethylenediamine]ruthenium(II), [(S,S)-N-(2-Amino-1,2-diphenylethyl)pentafluorobenzenesulphonamide]chloro(p-cymene)ruthenium(II), Chloro{[(1S,2S)-(-)-2-amino-1,2-diphenylethyl](pentafluorophenylsulphonyl)amido}(p-cymene)ruthenium(II)

R0122-200MG R0122-1G
TCIAR0122-200MGEA 54 EUR
TCIAR0122-200MG TCIAR0122-1G
RuCl[(S,S)-Fsdpen](p-cymene) ≥90% (by NMR)
RuCl[(S,S)-Fsdpen](p-cymene)
Formula: C₃₀H₂₈ClF₅N₂O₂RuS
Peso molecolare: 712,14 g/mol
MDL Number: MFCD12545954
CAS Number: 1026995-72-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=018a54109d7b0eea56041e53aeed0eae4fbdf145&vid=018a54109d7b0eea56041e53aeed0eae4fbdf145&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (19F-qNMR) min. 90 w/w%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR