Ritonavir ≥98.0% (by HPLC, total nitrogen)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
A-84538, ABT-538, Abbott 84538
R0116-200MG
R0116-1G
TCIAR0116-200MGEA
47
EUR
TCIAR0116-200MG
TCIAR0116-1G
Ritonavir ≥98.0% (by HPLC, total nitrogen)
Ritonavir
Formula:
C₃₇H₄₈N₆O₅S₂ Peso molecolare: 720,94 g/mol |
MDL Number:
MFCD00927142 CAS Number: 155213-67-5 Merck Index: 14,08238 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystaline |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 122.0 to 127.0 deg-C |
Specific rotation [a]20/D | +7.0 to +10.0 deg (C=2,MeOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...