Ranolazine dihydrochloride ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
Formula:
C₂₄H₃₃N₃O₄ |
MDL Number:
MFCD03788770 CAS Number: 95635-56-6 Merck Index: 14,08111 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...