Raloxifene hydrochloride ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
[6-Hydroxy-2-(4-hydroxyphenyl)benzo[b]thiophen-3-yl][4-[2-(1-piperidinyl)ethoxy]phenyl]methanone Hydrochloride
R0109-1G
R0109-5G
TCIAR0109-1GEA
169
EUR
TCIAR0109-1G
TCIAR0109-5G
Raloxifene hydrochloride ≥98.0% (by HPLC, titration analysis)
Raloxifene hydrochloride
Formula:
C₂₈H₂₇NO₄S Peso molecolare: 510,05 g/mol |
MDL Number:
MFCD01938233 CAS Number: 82640-04-8 Merck Index: 14,08098 |
Risultati del test di specificazione
Appearance | Light yellow to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...