Stampa…

Rosiglitazone ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: 5-(4-(2-(Methyl(pyridin-2-yl)amino)ethoxy)benzyl)thiazolidine-2,4-dione

R0106-200MG R0106-1G
TCIAR0106-200MGEA 19 EUR
TCIAR0106-200MG TCIAR0106-1G
Rosiglitazone ≥98.0% (by HPLC, titration analysis)
Rosiglitazone
Formula: C₁₈H₁₉N₃O₃S
Peso molecolare: 357,43 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00871760
CAS Number: 122320-73-4
Merck Index: 14,08265
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f54dcc0db2825f6fa36cd29221779d328902710c&vid=f54dcc0db2825f6fa36cd29221779d328902710c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 154.0 to 158.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR