Stampa…

Rotenone ≥95.0% (by HPLC)

Fornitore: TCI EUROPE
Pericolo

R0090-25G R0090-5G
TCIAR0090-25GEA 438 EUR
TCIAR0090-25G TCIAR0090-5G
Rotenone ≥95.0% (by HPLC)
Rotenone
Formula: C₂₃H₂₂O₆
Peso molecolare: 394,42 g/mol
Punto di ebollizione: 220°C
MDL Number: MFCD09025614
CAS Number: 83-79-4
EINECS: 201-501-9
UN: 2811
ADR: 6.1,II
Merck Index: 14,08271

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystaline
Purity (HPLC) min. 95.0 area%
Elemental analysis (Carbon) 67.00 to 71.00 %
Specific rotation [a]20/D -113.0 to -117.0 deg (C=1, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR