Stampa…

Rebamipide ≥98.0% (by HPLC)

Fornitore: TCI

R0085-1G R0085-25G R0085-5G
TCIAR0085-1GEA 29 EUR
TCIAR0085-1G TCIAR0085-25G TCIAR0085-5G
Rebamipide ≥98.0% (by HPLC)
Rebamipide
Formula: C₁₉H₁₅ClN₂O₄
Punto di fusione: 290°C
MDL Number: MFCD11114396
CAS Number: 90098-04-7
Merck Index: 14,08124
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0675f5a5df6fe982e0427fc79ad9f943a7ca02a&vid=114714f23a8c464b6b814cecb65f54b14f2167ac&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR