Butyl ricinoleate (by GC)
Fornitore: TCI EUROPE
Synonyms:
Ricinoleic Acid Butyl Ester, Ricinolic Acid Butyl Ester
R0045-25ML
R0045-500ML
TCIAR0045-25MLEA
16
EUR
TCIAR0045-25ML
TCIAR0045-500ML
Butyl ricinoleate (by GC)
Butyl ricinoleate
Formula:
C₂₂H₄₂O₃ Peso molecolare: 354,58 g/mol Punto di ebollizione: 275°C Flash Pt: 115°C |
MDL Number:
MFCD00136031 CAS Number: 151-13-3 EINECS: 205-785-5 |
Risultati del test di specificazione
Purity (GC) | min. 74.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...