Stampa…

Methyl ricinoleate ≥75.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Ricinolic Acid Methyl Ester, Ricinoleic Acid Methyl Ester

R0029-25ML R0029-500ML
TCIAR0029-25MLEA 16 EUR
TCIAR0029-25ML TCIAR0029-500ML
Methyl ricinoleate ≥75.0% (by GC)
Methyl ricinoleate
Formula: C₁₉H₃₆O₃
Peso molecolare: 312,49 g/mol
Flash Pt: 209°C
MDL Number: MFCD00046712
CAS Number: 141-24-2
EINECS: 205-472-3

Visualizza i codici

Risultati del test di specificazione

Purity (GC) min. 75.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR