Stampa…

Ricinoleic acid ≥80.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Ricinolic acid, (R)-12-Hydroxy-cis-9-octadecenoic acid, 12-Hydroxyoleic acid, Ricinelaidic acid, (R,Z)-12-Hydroxyoctadec-9-enoic acid

R0027-25ML R0027-500ML
TCIAR0027-25MLEA 17 EUR
TCIAR0027-25ML TCIAR0027-500ML
Ricinoleic acid ≥80.0% (by GC)
Ricinoleic acid
Formula: C₁₈H₃₄O₃
Peso molecolare: 298,47 g/mol
Flash Pt: 248°C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00084840
CAS Number: 141-22-0
EINECS: 205-470-2
Merck Index: 14,08211
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=56c0a0be6f65cd330808b00a009b8e909d044184&vid=56c0a0be6f65cd330808b00a009b8e909d044184&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Red to Green clear liquid to cloudy liquid
Purity (GC) min. 80.0 %
Purity (Neutralization titration) min. 85.0 %
Refractive index n20/D 1.4700 to 1.4730

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR