Stampa…

SQ 22536 ≥97.0% (by GC)

Fornitore: TCI

Synonyms: 9-(Tetrahydro-2-furyl)adenine, 9-(Tetrahydro-2-furanyl)-9H-purin-6-amine, 9-(Tetrahydrofuran-2-yl)-9H-purin-6-amine, SQ22536

Q0105-10MG Q0105-50MG
TCIAQ0105-10MGEA 101 EUR
TCIAQ0105-10MG TCIAQ0105-50MG
SQ 22536 ≥97.0% (by GC)
SQ 22536
Formula: C₉H₁₁N₅O
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00210216
CAS Number: 17318-31-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 162.0 to 167.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR