Stampa…

8-Quinolinecarbaldehyde ≥95.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Quinoline-8-carbaldehyde, 8-Formylquinoline, 8-Quinolinecarboxaldehyde, Quinoline-8-carboxaldehyde

Q0096-1G Q0096-5G
TCIAQ0096-1GEA 70 EUR
TCIAQ0096-1G TCIAQ0096-5G
8-Quinolinecarbaldehyde ≥95.0% (by GC)
8-Quinolinecarbaldehyde
Formula: C₁₀H₇NO
Peso molecolare: 157,17 g/mol
MDL Number: MFCD00805837
CAS Number: 38707-70-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0317b18cfbb70bf984ff6ceb6bae466a552174ce&vid=0317b18cfbb70bf984ff6ceb6bae466a552174ce&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 95.0 %
Melting point 92.0 to 97.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR