Stampa…

Quinoxalin-5-ol ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 5-Hydroxyquinoxaline

Q0095-200MG Q0095-1G
TCIAQ0095-200MGEA 18 EUR
TCIAQ0095-200MG TCIAQ0095-1G
Quinoxalin-5-ol ≥98.0% (by GC, titration analysis)
Quinoxalin-5-ol
Formula: C8H6N2O
Peso molecolare: 146,05 g/mol
Punto di ebollizione: 184°C
MDL Number: MFCD00972004
CAS Number: 17056-99-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dae473ffe791fe0eca6af4bd05912040513ab75b&vid=dae473ffe791fe0eca6af4bd05912040513ab75b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 99.0 to 103.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR